Merck
Lacinutrix chionocetis sp. nov., isolated from gut of a red snow crab.
Archives of microbiology (2016-12-29)
Hyangmi Kim, Sang-Chul Yoon, Kwang-Ho Choi, Sung-Tae Kim, Jae-Bong Lee, Dong-Sun Kim, Ho Le Han, Kyung Sook Bae, Doo-Sang Park
ABSTRACT
A Gram-negative, aerobic, non-motile, rod-shaped bacterial strain, designated MAB-07
Sigma-Aldrich
Bromosuccinic acid, 98%
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Gly-Asp, ≥99.0%
Sigma-Aldrich
Gly-Glu