Merck
PbSe quantum dot based luminescent solar concentrators.
Nanotechnology (2017-01-07)
Dennis L Waldron, Amanda Preske, Joseph M Zawodny, Todd D Krauss, Mool C Gupta
ABSTRACT
The results are presented for luminescent solar concentrators (LSCs) fabricated with poly(lauryl methacrylate-co-ethylene glycol dimethacrylate) (P(LMA-co-EGDMA)) and Angstrom Bond, Inc. AB9093 acrylic epoxy matrix, high quantum yield (> 70%) PbSe quantum dots (QDs) and silicon photovoltaic (Si PV) cells. LSCs were tested under a lamp with broadband illumination, photon flux-matched to a standard solar spectrum and verified under a calibrated solar lamp source. The P(LMA-co-EGDMA) sample demonstrated the highest power conversion efficiency of any known LSC fabricated with either QDs or Si PV cells, 4.74%. Additionally, increased temperature was shown to reduce efficiency.
Sigma-Aldrich
Lauryl methacrylate, contains 500 ppm MEHQ as inhibitor, 96%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor