Merck
  • Bisphenol A and its analogues disrupt centrosome cycle and microtubule dynamics in prostate cancer.
Bisphenol A and its analogues disrupt centrosome cycle and microtubule dynamics in prostate cancer.
Endocrine-related cancer (2016-12-22)
Shuk-Mei Ho, Rahul Rao, Sarah To, Emma Schoch, Pheruza Tarapore
ABSTRACT
Humans are increasingly exposed to structural analogues of bisphenol A (BPA), as BPA is being replaced by these compounds in BPA-free consumer products. We have previously shown that chronic and developmental exposure to BPA is associated with increased prostate cancer (PCa) risk in human and animal models. Here, we examine whether exposure of PCa cells (LNCaP, C4-2) to low-dose BPA and its structural analogues (BPS, BPF, BPAF, TBBPA, DMBPA and TMBPA) affects centrosome amplification (CA), a hallmark of cancer initiation and progression. We found that exposure to BPA, BPS, DMBPA and TBBPA, in descending order, increased the number of cells with CA, in a non-monotonic dose-response manner. Furthermore, cells treated with BPA and their analogues initiated centrosome duplication at 8 h after release from serum starvation, significantly earlier in G-1 phase than control cells. This response was attended by earlier release of nucleophosmin from unduplicated centrosomes. BPA-exposed cells exhibited increased expression of cyclin-dependent kinase CDK6 and decreased expression of CDK inhibitors (p21Waf1/CIP1 and p27KIP1). Using specific antagonists for estrogen/androgen receptors, CA in the presence of BPA or its analogues was likely to be mediated via ESR1 signaling. Change in microtubule dynamics was observed on exposure to these analogues, which, for BPA, was accompanied by increased expression of centrosome-associated protein CEP350 Similar to BPA, chronic treatment of cells with DMBPA, but not other analogues, resulted in the enhancement of anchorage-independent growth. We thus conclude that selected BPA analogues, similar to BPA, disrupt centrosome function and microtubule organization, with DMBPA displaying the broadest spectrum of cancer-promoting effects.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-PCNT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1