Merck
  • TFEB and TFE3 cooperate in the regulation of the innate immune response in activated macrophages.
TFEB and TFE3 cooperate in the regulation of the innate immune response in activated macrophages.
Autophagy (2016-05-14)
Nunzia Pastore, Owen A Brady, Heba I Diab, José A Martina, Lu Sun, Tuong Huynh, Jeong-A Lim, Hossein Zare, Nina Raben, Andrea Ballabio, Rosa Puertollano
ABSTRACT
The activation of transcription factors is critical to ensure an effective defense against pathogens. In this study we identify a critical and complementary role of the transcription factors TFEB and TFE3 in innate immune response. By using a combination of chromatin immunoprecipitation, CRISPR-Cas9-mediated genome-editing technology, and in vivo models, we determined that TFEB and TFE3 collaborate with each other in activated macrophages and microglia to promote efficient autophagy induction, increased lysosomal biogenesis, and transcriptional upregulation of numerous proinflammatory cytokines. Furthermore, secretion of key mediators of the inflammatory response (CSF2, IL1B, IL2, and IL27), macrophage differentiation (CSF1), and macrophage infiltration and migration to sites of inflammation (CCL2) was significantly reduced in TFEB and TFE3 deficient cells. These new insights provide us with a deeper understanding of the transcriptional regulation of the innate immune response.
Sigma-Aldrich
Anti-TFE3 antibody, Mouse monoclonal, clone TFE3-37, purified from hybridoma cell culture
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, BioXtra, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution