Merck
  • New synthesis of vaulted biaryl ligands via the Snieckus phenol synthesis.
New synthesis of vaulted biaryl ligands via the Snieckus phenol synthesis.
Organic letters (2005-01-28)
Su Yu, Constantinos Rabalakos, William D Mitchell, William D Wulff
ABSTRACT
[reaction: see text] In an effort to develop a synthesis of the VAPOL ligand that avoids the use of a chromium carbene complex, a route was examined that involved the annulation of a naphthalene carboxamide via the method of Snieckus. The latter derivatives could be converted in a two-step sequence to 2-phenyl-4-phenanthrols in 60-72% overall yields. The utility of this method for the synthesis of VAPOL derivatives is demonstrated in the synthesis of (S)-7,7'-dimethyl-VAPOL.
Sigma-Aldrich
(S)-VANOL, 97%
Sigma-Aldrich
(R)-VANOL, 97%
Sigma-Aldrich
(S)-VAPOL, 97%
Sigma-Aldrich
(R)-VAPOL, 97%