Merck
  • Copper-diamine-catalyzed N-arylation of pyrroles, pyrazoles, indazoles, imidazoles, and triazoles.
Copper-diamine-catalyzed N-arylation of pyrroles, pyrazoles, indazoles, imidazoles, and triazoles.
The Journal of organic chemistry (2004-08-17)
Jon C Antilla, Jeremy M Baskin, Timothy E Barder, Stephen L Buchwald
ABSTRACT
This paper details the copper-catalyzed N-arylation of pi-excessive nitrogen heterocycles. The coupling of either aryl iodides or aryl bromides with common nitrogen heterocycles (pyrroles, pyrazoles, indazoles, imidazoles, and triazoles) was successfully performed in good yield with catalysts derived from diamine ligands and CuI. General conditions were found that tolerate functional groups such as aldehydes, ketones, alcohols, primary amines, and nitriles on the aryl halide or heterocycle. Hindered aryl halides or heterocycles were also found to be suitable substrates using the conditions reported herein.
Sigma-Aldrich
3-Chloroindazole, 97%
Sigma-Aldrich
2-Ethylpyrrole, technical grade, 90%