Merck
  • AFM forces measured between gold surfaces coated with self-assembled monolayers of 1-hexadecanethiol.
AFM forces measured between gold surfaces coated with self-assembled monolayers of 1-hexadecanethiol.
Langmuir : the ACS journal of surfaces and colloids (2008-06-26)
Jialin Wang, Roe-Hoan Yoon
ABSTRACT
An atomic force microscope (AFM) was used to measure the forces between gold surfaces with and without hydrophobizing them by the self-assembly of 1-hexadecanethiol. The forces measured between bare gold surfaces were fitted to the Derjaguin-Landau-Verwey-Overbeek (DLVO) theory with a Hamaker constant of 1.2 x 10 (-20) J, which was close to the value determined using the methylene iodide contact angle method but was lower than that calculated using the Lifshitz theory. When the surfaces were hydrophobized in a 0.01 mM thiol-in-ethanol solution for 10 min, the measured forces exhibited a long-range force with a decay length of 35 nm. Despite its high water contact angle (105 degrees ), the force curve was smooth and exhibited no steps. When the surfaces were hydrophobized in a 1 mM thiol solution for longer than 6 h, however, the force curves exhibited steps, indicating that the long-range attractions were caused by bridging bubbles. When the measurements were conducted after washing the substrates with organic solvents, the steps disappeared and long-range attractive forces appeared. In the presence of ethanol, the water contact angle decreased to below 90 degrees , the attraction became weaker, and the force curves became smooth. On the basis of the results obtained in the present work, possible mechanisms for the long-range attractions are discussed.
Sigma-Aldrich
1-Hexadecanethiol, 99%
Sigma-Aldrich
1-Hexadecanethiol, ≥95.0% (GC)