Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Potassium disulfite, BioUltra, ≥97.0% (RT)
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, puriss., 99.0-101.0% (calc. on H2O free substance), meets analytical specification of Ph. Eur., BP, USP, FC, E 337
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (perchloric acid titration)
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, ACS reagent, 99%
Sigma-Aldrich
Potassium disulfite, ≥98%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.5%
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis