Merck
  • Erythropoietin ameliorates renal interstitial fibrosis via the inhibition of fibrocyte accumulation.
Erythropoietin ameliorates renal interstitial fibrosis via the inhibition of fibrocyte accumulation.
Molecular medicine reports (2015-01-13)
Xu Chang Geng, Zhou Pang Hu, Guo Yong Lian
ABSTRACT
Erythropoietin (EPO) is a hematopoietic hormone that protects against renal interstitial fibrosis in animal models; however, the mechanism underlying the anti‑fibrotic activity of EPO has remained elusive. The present study aimed to elucidate this mechanism. Twenty‑four male C57BL6 mice were randomly divided into four groups, each comprising six mice: (i) control group (Sh); (ii) unilateral ureteral obstruction (UUO) plus vehicle group (U+V); (ⅲ) UUO plus 300 U/kg body weight recombinant human (rh)EPO (U+E1) and (ⅳ) UUO plus 1,000 U/kg body weight rhEPO (U+E2). Seven days post‑surgery, the mice were sacrificed for examination. UUO induced significant deposition of extracellular matrix, detected by picro‑sirius red staining, which was decreased following rhEPO treatment. UUO also induced deposition of collagen I and fibronectin, rhEPO treatment was able to attenuate this effect at protein and mRNA levels. Compared with the control groups, UUO resulted in the accumulation of α‑smooth muscle actin‑positive cells in the interstitium, an effect which was ameliorated by rhEPO. Furthermore, rhEPO abrogated the UUO‑induced increase in the number of bone marrow‑derived myofibroblasts. Mechanistically, it was discovered that rhEPO decreased CXC chemokine ligand 16 (CXCL16) expression at protein level. However, treatment with rhEPO did not alter the protein expression of CC chemokine ligand 21 or CXCL12. These results suggested that rhEPO decreased fibrocyte accumulation via the suppression of renal CXCL16, which resulted in the attenuation of renal fibrosis.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cxcl12
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cxcl16
Sigma-Aldrich
MISSION® esiRNA, targeting human FN1
Sigma-Aldrich
CXCL16 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprc
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission