Merck
  • Gene Cloning, High-Level Expression, and Characterization of an Alkaline and Thermostable Lipase from Trichosporon coremiiforme V3.
Gene Cloning, High-Level Expression, and Characterization of an Alkaline and Thermostable Lipase from Trichosporon coremiiforme V3.
Journal of microbiology and biotechnology (2015-01-16)
Jian-Rong Wang, Yang-Yuan Li, Danni Liu
ABSTRACT
The present study describes the gene cloning and high-level expression of an alkaline and thermostable lipase gene from Trichosporon coremiiforme V3. Nucleotide analysis revealed that this lipase gene has an open reading frame of 1,692 bp without any introns, encoding a protein of 563 amino acid residues. The lipase gene without its signal sequence was cloned into plasmid pPICZαA and overexpressed in Pichia pastoris X33. The maximum lipase activity of recombinant lipase was 5,000 U/ml, which was obtained in fed-batch cultivation after 168 h induction with methanol in a 50 L bioreactor. The purified lipase showed high temperature tolerance, and being stable at 60 °C and kept 45% enzyme activity after 1 h incubation at 70 °C. The stability, effects of metal ions and other reagents were also determined. The chain length specificity of the recombinant lipase showed high activity toward triolein (C18:1) and tripalmitin (C16:0).
Sigma-Aldrich
Methyl stearate, ≥96%, FG
Sigma-Aldrich
Methyl palmitate, ≥97%
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl stearate, ~99% (GC)
Sigma-Aldrich
Methyl butyrate, 99%
Sigma-Aldrich
Methyl butyrate, ≥98%, FG
Sigma-Aldrich
Methyl butyrate, natural, ≥98%, FG
Sigma-Aldrich
Methyl laurate, ≥98%, FG
Sigma-Aldrich
Methyl laurate, 99.5%