Merck
  • Interstitial deletion at 11q14.2-11q22.1 may cause severe learning difficulties, mental retardation and mild heart defects in 13-year old male.
Interstitial deletion at 11q14.2-11q22.1 may cause severe learning difficulties, mental retardation and mild heart defects in 13-year old male.
Molecular cytogenetics (2015-09-22)
Ioannis Papoulidis, Vassilis Paspaliaris, Elisavet Siomou, Sandro Orru, Roberta Murru, Stavros Sifakis, Petros Nikolaidis, Antonios Garas, Sotirios Sotiriou, Loretta Thomaidis, Emmanouil Manolakos
ABSTRACT
Interstitial deletions of the long arm of chromosome 11 are rare, and they could be assumed as non-recurrent chromosomal rearrangements due to high variability of the size and the breakpoints of the deleted region. The exact region of the deletion was difficult to be determined before the use of molecular cytogenetic techniques such as array comparative genomic hybridization (aCGH). Here, a 13-year old boy with severe learning difficulties, mental retardation and mild heart defects is described. Conventional G-band karyotyping was performed and it is found that the patient is a carrier of a de novo interstitial deletion on the long arm of chromosome 11, involving 11q14 and 11q22 breakpoints. Further investigation, using aCGH, specified the deleted region to 11q14.2-11q22.1. There was a difficulty in correlating the genotype with the phenotype of the patient due to lack of similar cases in literature. More studies should be done in order to understand the genetic background that underlies the phenotypic differences observed in similar cases.
Sigma-Aldrich
EED human, recombinant, expressed in baculovirus infected insect cells, ≥90% (SDS-PAGE)