Merck
  • Reciprocal activation of CD4+ T cells and synovial fibroblasts by stromal cell-derived factor 1 promotes RANKL expression and osteoclastogenesis in rheumatoid arthritis.
Reciprocal activation of CD4+ T cells and synovial fibroblasts by stromal cell-derived factor 1 promotes RANKL expression and osteoclastogenesis in rheumatoid arthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2014-02-28)
Hae-Rim Kim, Kyoung-Woon Kim, Bo-Mi Kim, Hong-Geun Jung, Mi-La Cho, Sang-Heon Lee
ABSTRACT
Stromal cell-derived factor 1 (SDF-1) is a chemokine that is involved in the bone-destructive process in rheumatoid arthritis (RA) and bony metastasis in malignancy. This study was undertaken to determine the role and mechanism of SDF-1 in RA-associated osteoclastogenesis. The expression of SDF-1, tumor necrosis factor α (TNFα), and RANKL in RA synovial tissue was analyzed using confocal microscopy. After synovial fibroblasts and CD4+ T cells were treated with SDF-1, RANKL messenger RNA expression was determined by real-time and reverse transcription polymerase chain reaction. Osteoclastogenesis was assessed by counting tartrate-resistant acid phosphatase-positive multinucleated cells in CD14+ monocytes cultured with SDF-1 in the presence of anticytokine antibodies or signal inhibitors and in monocytes cocultured with SDF-1-pretreated synovial fibroblasts and CD4+ T cells. RANKL, TNFα, and SDF-1 were coexpressed in the lining and sublining of RA synovium. SDF-1 stimulated RANKL expression in RA synovial fibroblasts and CD4+ T cells, and TNFα inhibition reduced this stimulation. When monocytes isolated from human peripheral blood were cultured with SDF-1, they were differentiated into osteoclasts in the absence of RANKL. Monocytes were also differentiated into osteoclasts when they were cocultured with SDF-1-pretreated synovial fibroblasts or CD4+T cells; however, this osteoclastogenesis was reduced by TNFα inhibition. Our findings indicate that SDF-1 induces osteoclastogenesis directly and indirectly via up-regulating RANKL expression in RA synovial fibroblasts and CD4+ T cells, and that this is mediated by TNFα. The axis of SDF-1 and RANKL is a potential therapeutic target for RA-associated bone destruction.
Product Description
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material