Merck
  • Fabrication of fluorescent SiO2@zeolitic imidazolate framework-8 nanosensor for Cu(2+) detection.
Fabrication of fluorescent SiO2@zeolitic imidazolate framework-8 nanosensor for Cu(2+) detection.
The Analyst (2014-11-27)
Yonghai Song, Dongqin Hu, Fenfen Liu, Shouhui Chen, Li Wang
ABSTRACT
A simple strategy to fabricate a fluorescent SiO2@zeolitic imidazolate framework-8 (ZIF-8) core-shell nanosensor for Cu(2+) detection was demonstrated in this work. The nanosensor was synthesized using carboxyl-functionalized SiO2 nanoparticles (SiO2 NPs) as a template to induce the growth of ZIF-8 on its surface. The porous SiO2@ZIF-8 exhibited extremely good adsorption properties and a large specific surface area to accumulate Cu(2+), and the pyridyl nitrogen sites in imidazole played vital roles in the recognition of Cu(2+). The fluorescent intensity decreased linearly with the increasing of Cu(2+) concentration in the range of 10-500 nM and the detection limit was estimated to be 3.8 nM. The SiO2@ZIF-8 nanosensor could be further used to determine trace amounts of Cu(2+) in real water samples, while some previous sensors had to be dispersed in organic solution for use, such as DMSO and MeCN. The core-shell nanostructures of SiO2@ZIF-8 made it possible for it to be dispersed directly in aqueous solution and prevented ZIF-8 from aggregation, which enhanced the sensing performance of the SiO2@ZIF-8 nanosensor.
Product Description
Ondansetron impurity F, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
2-Methylimidazole, 99%
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%