Merck
  • Identification of glutathione S-transferase omega 1 (GSTO1) protein as a novel tumor-associated antigen and its autoantibody in human esophageal squamous cell carcinoma.
Identification of glutathione S-transferase omega 1 (GSTO1) protein as a novel tumor-associated antigen and its autoantibody in human esophageal squamous cell carcinoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-03)
Yang Li, Qi Zhang, Bo Peng, Qing Shao, Wei Qian, Jian-Ying Zhang
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is the main form of esophageal malignancy. The approach for early diagnosis of this malignancy is very limited. In the present study, we first evaluated glutathione S-transferase omega 1 (GSTO1), a protein related to metabolism, as a tumor-associated antigen in ESCC, and we also evaluated its autoantibody as a potential biomarker in early detection of ESCC. First, immunohistochemistry (IHC) analysis of GSTO1 protein expression in esophageal tissues showed that the percentage of positive staining of GSTO1 in ESCC tissues was 87.5% while there was no positive staining in adjacent tissues or normal tissues, indicating that overexpression of GSTO1 is closely related to ESCC. Then, enzyme-linked immunosorbent assay (ELISA) showed that the frequency of detectable autoantibody against GSTO1 in patients' sera totals 44.8%. In contrast, the frequency of detectable autoantibody was only 6.7% in normal human sera (p < 0.01). To further evaluate our ELISA results, western blotting and immunofluorescence assay were also performed. The results were consistent with the data from ELISA. In conclusion, the current study has demonstrated that GSTO1 protein is overexpressed in ESCC and can induce a detectable autoantibody response, which may serve as a potential biomarker in the early detection of ESCC.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
GST O1-1, Recombinant Human
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)