Merck
  • Phase II study of perifosine and sorafenib dual-targeted therapy in patients with relapsed or refractory lymphoproliferative diseases.
Phase II study of perifosine and sorafenib dual-targeted therapy in patients with relapsed or refractory lymphoproliferative diseases.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-09-23)
Anna Guidetti, Carmelo Carlo-Stella, Silvia L Locatelli, Walter Malorni, Roberta Mortarini, Simonetta Viviani, Domenico Russo, Alfonso Marchianò, Roberto Sorasio, Anna Dodero, Lucia Farina, Laura Giordano, Massimo Di Nicola, Andrea Anichini, Paolo Corradini, Alessandro M Gianni
ABSTRACT
To evaluate safety and activity of perifosine and sorafenib combination therapy in patients with lymphoproliferative diseases. Patients with relapsed and refractory lymphoproliferative diseases received perifosine (50 mg twice daily) for 1 month. Patients achieving less than partial response (PR) after perifosine alone were administered the combination therapy [perifosine plus sorafenib (400 mg twice daily)] until progressive disease (PD) or unacceptable toxicity occurred. The pERK and pAKT in peripheral blood lymphocytes as well as serum cytokine levels were investigated as predictive biomarkers of response. Forty patients enrolled in this study. After 1 month of perifosine alone, 36 who achieved less than PR went on to combination therapy, whereas four patients with chronic lymphocytic leukemia (CLL) who achieved PR continued with perifosine alone for a median of 10 months (range, 4-21). The most common drug-related toxicities were grade 1-2 anemia (17%), thrombocytopenia (9%), diarrhea (25%), joint pain (22%), and hand-foot skin reaction (25%). Three patients experienced grade 3 pneumonitis. Eight patients (22%) achieved PR, 15 (42%) achieved stable disease, and 13 (36%) experienced PD. A 28% PR rate was recorded for 25 patients with Hodgkin lymphoma. Among all patients, median overall survival and progression-free survival were 16 and 5 months, respectively. Early reductions in pERK and pAKT significantly correlated with the probability of clinical response. Perifosine and sorafenib combination therapy is feasible with manageable toxicity and demonstrates promising activity in patients with Hodgkin lymphoma. The predictive value of pERK and pAKT should be confirmed in a larger patient cohort.
Sigma-Aldrich
Human TARC / CCL17 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder