Merck
  • Technical Brief: a novel strategy for enrichment of trabecular meshwork protease proteome.
Technical Brief: a novel strategy for enrichment of trabecular meshwork protease proteome.
Molecular vision (2008-05-21)
Renata Picciani, Anna K Junk, Sanjoy K Bhattacharya
ABSTRACT
We present a novel and simple enrichment strategy to capture trabecular meshwork (TM) protease proteome. The method relies on fractionation of TM tissue into cytosolic and nuclear extracts and subsequent affinity enrichment of proteases on peptide inhibitors. A large repertoire of available protease substrate analog peptides enables an improved capture of TM protease proteome compared to SDS-PAGE fractionation alone. Peptide analog inhibitors of protease substrates are immobilized on a protein A or G column using 254 nm intense ultraviolet (UV) light. The TM cytosolic protein extract incubated on the column is eluted with salt or a buffer with a low hydrogen ion concentration. The resultant protein solution is precipitated with acetone, fractionated on SDS-PAGE, in situ trypsin digested, and subjected to mass spectrometry. This relatively simple protocol enables improved capture of cytosolic proteases. We identified 20 previously reported TM proteins from a single donor tissue using affinity enrichment. The majority of identified proteins were either intracellular proteases or known protease inhibitors. Both serine and cysteine proteases were captured using this strategy with improved coverage compared to our previous identification without affinity enrichment.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, clone A3S, Upstate®, from rabbit
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution