Merck
  • NURD, a novel complex with both ATP-dependent chromatin-remodeling and histone deacetylase activities.
NURD, a novel complex with both ATP-dependent chromatin-remodeling and histone deacetylase activities.
Molecular cell (1999-01-14)
Y Xue, J Wong, G T Moreno, M K Young, J Côté, W Wang
ABSTRACT
ATP-dependent chromatin-remodeling complexes are known to facilitate transcriptional activation by opening chromatin structures. We report a novel human complex, named NURD, which contains not only ATP-dependent nucleosome disruption activity, but also histone deacetylase activity, which usually associates with transcriptional repression. The deacetylation is stimulated by ATP on nucleosomal templates, suggesting that nucleosome disruption aids the deacetylase to access its substrates. One subunit of NURD was identified as MTA1, a metastasis-associated protein with a region similar to the nuclear receptor core-pressor, N-CoR; and antibodies against NURD partially relieve transcriptional repression by thyroid hormone receptor. These results suggest that ATP-dependent chromatin remodeling can participate in transcriptional repression by assisting repressors in gaining access to chromatin.