Merck
Binding of ArgTX-636 in the NMDA receptor ion channel.
Journal of molecular biology (2014-05-28)
Mette H Poulsen, Jacob Andersen, Rune Christensen, Kasper B Hansen, Stephen F Traynelis, Kristian Strømgaard, Anders Skov Kristensen
ABSTRACT
The N-methyl-d-aspartate receptors (NMDARs) constitute an important class of ligand-gated cation channels that are involved in the majority of excitatory neurotransmission in the human brain. Compounds that bind in the NMDAR ion channel and act as blockers are use- and voltage-dependent inhibitors of NMDAR activity and have therapeutic potential for treatment of a variety of brain diseases or as pharmacological tools for studies of the neurobiological role of NMDARs. We have performed a kinetic analysis of the blocking mechanism of the prototypical polyamine toxin NMDAR ion channel blocker argiotoxin-636 (ArgTX-636) at recombinant GluN1/2A receptors to provide detailed information on the mechanism of block. The predicted binding site of ArgTX-636 is in the pore region of the NMDAR ion channel formed by residues in the transmembrane M3 and the M2 pore-loop segments of the GluN1 and GluN2A subunits. To assess the predicted binding mode in further detail, we performed an alanine- and glycine-scanning mutational analysis of this pore-loop segment to systematically probe the role of pore-lining M2 residues in GluN1 and GluN2A in the channel block by ArgTX-636. Comparison of M2 positions in GluN1 and GluN2A where mutation influences ArgTX-636 potency suggests differential contribution of the M2-loops of GluN1 and GluN2A to binding of ArgTX-636. The results of the mutational analysis are highly relevant for the future structure-based development of argiotoxin-derived NMDAR channel blockers.
Sigma-Aldrich
L-Glutamic acid hydrochloride
Pricing and availability is not currently available.
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Pricing and availability is not currently available.
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.