Merck
  • Stereoisomer analysis of wastewater-derived beta-blockers, selective serotonin re-uptake inhibitors, and salbutamol by high-performance liquid chromatography-tandem mass spectrometry.
Stereoisomer analysis of wastewater-derived beta-blockers, selective serotonin re-uptake inhibitors, and salbutamol by high-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2007-10-05)
Sherri L MacLeod, Priya Sudhir, Charles S Wong
ABSTRACT
A reversed-phase enantioselective liquid chromatography-tandem mass spectrometry (HPLC-MS-MS) method was developed to measure enantiomer fractions (EF) and concentrations of pharmaceuticals in wastewater. Enantiomer resolution of six beta-blockers (atenolol, metoprolol, nadolol, pindolol, propranolol, and sotalol) along with two selective serotonin re-uptake inhibitors (citalopram, fluoxetine) and one beta(2)-agonist (salbutamol) was achieved with the Chirobiotic V stationary phase. Analyte recovery averaged 86% in influent and 78% in effluent with limits of detection ranging from 0.2 to 7.5 ng/L. These results represent an improvement in wastewater EF measurement for atenolol, metoprolol and propranolol as well as the first EF measurements of citalopram, fluoxetine, nadolol, pindolol, salbutamol and sotalol in wastewaters. Changes in EF through treatment indicate biologically mediated stereoselective processes were likely occurring during wastewater treatment.
Product Description
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Sigma-Aldrich
(S)-(−)-Atenolol, powder