Merck
  • T-2 toxin, a trichothecene mycotoxin: review of toxicity, metabolism, and analytical methods.
T-2 toxin, a trichothecene mycotoxin: review of toxicity, metabolism, and analytical methods.
Journal of agricultural and food chemistry (2011-03-23)
Yanshen Li, Zhanhui Wang, Ross C Beier, Jianzhong Shen, David De Smet, Sarah De Saeger, Suxia Zhang
ABSTRACT
This review focuses on the toxicity and metabolism of T-2 toxin and analytical methods used for the determination of T-2 toxin. Among the naturally occurring trichothecenes in food and feed, T-2 toxin is a cytotoxic fungal secondary metabolite produced by various species of Fusarium. Following ingestion, T-2 toxin causes acute and chronic toxicity and induces apoptosis in the immune system and fetal tissues. T-2 toxin is usually metabolized and eliminated after ingestion, yielding more than 20 metabolites. Consequently, there is a possibility of human consumption of animal products contaminated with T-2 toxin and its metabolites. Several methods for the determination of T-2 toxin based on traditional chromatographic, immunoassay, or mass spectroscopy techniques are described. This review will contribute to a better understanding of T-2 toxin exposure in animals and humans and T-2 toxin metabolism, toxicity, and analytical methods, which may be useful in risk assessment and control of T-2 toxin exposure.
Product Description
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Pricing and availability is not currently available.