Merck
  • A comprehensive review on vanilla flavor: extraction, isolation and quantification of vanillin and others constituents.
A comprehensive review on vanilla flavor: extraction, isolation and quantification of vanillin and others constituents.
International journal of food sciences and nutrition (2007-09-22)
Arun K Sinha, Upendra K Sharma, Nandini Sharma
ABSTRACT
Vanilla, being the world's most popular flavoring materials, finds extensive applications in food, beverages, perfumery and pharmaceutical industry. With the high demand and limited supply of vanilla pods and the continuing increase in their cost, numerous efforts of blending and adulteration in natural vanilla extracts have been reported. Thus, to ensure the quality of vanilla extracts and vanilla-containing products, it is important to develop techniques to verify their authenticity. Quantitatively, vanillin is the major compound present in the vanilla pods and the determination of vanillin is a vital consideration in natural vanilla extracts. This paper provides a comprehensive account of different extraction processes and chromatographic techniques applied for the separation, identification and determination of chemical constituents of vanilla. The review also provides an account of different methods applied for the quantification and the authentification of chemical constituents of vanilla extract. As the various properties of vanilla are attributed to its main constituent vanillin, its physico-chemical and bioactive properties have also been outlined.
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Vanillin, European Pharmacopoeia (EP) Reference Standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)