Merck
  • Is primidone the drug of choice for epileptic patients with QT-prolongation? A comprehensive analysis of literature.
Is primidone the drug of choice for epileptic patients with QT-prolongation? A comprehensive analysis of literature.
Seizure (2005-11-29)
D Christidis, D Kalogerakis, T Y Chan, D Mauri, G Alexiou, A Terzoudi
ABSTRACT
Sudden unexplained/unexpected death (SUDEP) in epilepsy is a major cause of death accounting for 7-17% of the mortality among epileptic patients. Prolongation of QT-interval has been issued as a major mechanism in SUDEP since it is associated with fatal cardiac arrhythmias. This condition may be further precipitated by anti-epileptic treatment. Despite thorough literature research, we did not find any reports suggesting that primidone is responsible for QT-prolongation. On the contrary, all the retrieved reports addressed that the drug shortened QT-interval and corrected signs and symptoms of the underlying disease.
Product Description
Supelco
Primidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Primidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Primidone, analytical standard