Merck
  • Fulvestrant, a selective estrogen receptor down-regulator, sensitizes estrogen receptor negative breast tumors to chemotherapy.
Fulvestrant, a selective estrogen receptor down-regulator, sensitizes estrogen receptor negative breast tumors to chemotherapy.
Cancer letters (2014-01-28)
Donghai Jiang, Yuan Huang, Ning Han, Mingjie Xu, Liang Xu, Lin Zhou, Shu Wang, Weimin Fan
ABSTRACT
Drug resistance frequently results in poor prognosis and high 5-year recurrence rate in estrogen receptor-negative (ER-) breast cancer patients. Herein, we examined the reversal effects of fulvestrant on multidrug resistance (MDR) in ER- breast cancer cells. Co-administration of fulvestrant significantly sensitized ER- MDR tumors to paclitaxel both in vitro and in vivo. Further analyses indicated that fulvestrant did not affect P-gp expression, but could inhibit P-gp function and subsequently reverse P-gp mediated drug resistance in ER- breast cancer cells. These results showed that combination of fulvestrant and chemotherapeutic agents might provide an effective treatment for ER- MDR breast cancers.
Product Description
Fulvestrant, European Pharmacopoeia (EP) Reference Standard
Fulvestrant for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fulvestrant, >98% (HPLC)