Merck
  • In situ nitrogenated graphene-few-layer WS2 composites for fast and reversible Li+ storage.
In situ nitrogenated graphene-few-layer WS2 composites for fast and reversible Li+ storage.
Nanoscale (2013-07-16)
Dongyun Chen, Ge Ji, Bo Ding, Yue Ma, Baihua Qu, Weixiang Chen, Jim Yang Lee
ABSTRACT
Two-dimensional nanosheets can leverage on their open architecture to support facile insertion and removal of Li(+) as lithium-ion battery electrode materials. In this study, two two-dimensional nanosheets with complementary functions, namely nitrogen-doped graphene and few-layer WS2, were integrated via a facile surfactant-assisted synthesis under hydrothermal conditions. The layer structure and morphology of the composites were confirmed by X-ray diffraction, scanning electron microscopy and high-resolution transmission microscopy. The effects of surfactant amount on the WS2 layer number were investigated and the performance of the layered composites as high energy density lithium-ion battery anodes was evaluated. The composite formed with a surfactant : tungsten precursor ratio of 1 : 1 delivered the best cyclability (average of only 0.08% capacity fade per cycle for 100 cycles) and good rate performance (80% capacity retention with a 50-fold increase in current density from 100 mA g(-1) to 5000 mA g(-1)), and may find uses in power-oriented applications.
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis