Merck
  • Regulation of rabbit cytochrome P450 2E1 expression in HepG2 cells by insulin and thyroid hormone.
Regulation of rabbit cytochrome P450 2E1 expression in HepG2 cells by insulin and thyroid hormone.
Molecular pharmacology (1998-10-10)
H M Peng, M J Coon
ABSTRACT
The regulation of cytochrome P450 (CYP) 2E1, the ethanol-inducible isoform, is particularly complex. The level is affected by a variety of other foreign compounds, by insulin (as studied in several laboratories), and by triiodothyronine (T3), which has not been previously examined at the molecular level. In the present investigation, a stably transfected HepG2 cell line harboring a rabbit CYP2E1 minigene containing the coding sequence together with 1.6 kilobases of the 5' flanking region and the untranslated region (UTR), as well as 0.5 kilobases of the 3' UTR, was established. Western blot analysis showed that 1 microM insulin decreased the CYP2E1 protein level in a dose- and time-dependent manner, whereas 1 microM T3 increased the level 2-fold in 1 day and 8-fold in 5 days. Similarly, steady state CYP2E1 mRNA levels were decreased by insulin but were increased by T3. Neither hormone affected the transcription rate of the CYP2E1 5' flanking region with an UTR/luciferase fusion gene, indicating that the regulation is post-transcriptional in this system under our experimental conditions. When the CYP2E1 3' UTR was removed from the minigene, CYP2E1 mRNA and protein were up-regulated by insulin but were not affected by T3. These findings, including mRNA half-life determinations, indicate that the effects of insulin and T3 are a result of altered mRNA stability and that the 3' UTR of CYP2E1 contains regulatory information for these hormone-mediated effects.
Product Description
ECL Direct Nucleic Acid, Cytiva RPN3000, pack of 1 ea
ECL Detection Reagents, Cytiva RPN3004, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3005, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3001, pack of 1 ea