- Distribution of trimipramine and its major metabolites in a fatal overdose case.
Distribution of trimipramine and its major metabolites in a fatal overdose case.
Journal of analytical toxicology (1987-07-01)
A D Fraser, A F Isner, R A Perry
PMID3626529
ABSTRACT
A fatality following ingestion of the tricyclic antidepressant trimipramine (Surmontil) is described. Quantitation was performed by high-performance liquid chromatography. Trimipramine and desmethyltrimipramine concentrations were 4.8 and 2.1 mg/L, respectively, in postmortem blood. The concentration of trimipramine, desmethyltrimipramine, and their respective 2-hydroxy metabolites were also measured in liver and urine. Analysis of gastric contents revealed a tricyclic antidepressant drug. These findings are compared to previously published reports of trimipramine-related fatalities.
Product Description
Supelco
N-Desmethyltrimipramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®