Merck
Role of drug therapy for sustained ventricular tachyarrhythmias.
Cardiology clinics (2008-06-10)
L Brent Mitchell
ABSTRACT
Antiarrhythmic drug therapy, broadly defined, is the mainstay of treatment and prevention of ventricular tachycardia (VT)/ventricular fibrillation (VF), which can lead to sudden death. This article evaluates the evidence for and appropriate use of class I antiarrhythmic drugs, class III antiarrhythmic drugs, beta-blockers, nondihydropyridine calcium-channel blockers, statins, angiotensin enzyme inhibitors, angiotensin receptor blockers, aldosterone blockers, and digoxin for antiarrhythmic benefits in patients who have a propensity for VT/VF and therefore are at risk of sudden death.
Sigma-Aldrich
Azimilide dihydrochloride, ≥97% (HPLC)