Merck
  • Macromolecular prodrugs of ribavirin combat side effects and toxicity with no loss of activity of the drug.
Macromolecular prodrugs of ribavirin combat side effects and toxicity with no loss of activity of the drug.
Chemical communications (Cambridge, England) (2013-02-23)
Mille B L Kryger, Benjamin M Wohl, Anton A A Smith, Alexander N Zelikin
ABSTRACT
Chemi-enzymatic synthesis of ribavirin acrylate and subsequent RAFT co-polymerization with acrylic acid afforded a formulation of a broad spectrum antiviral drug which avoids accumulation in erythrocytes, the origin of the main side effect of ribavirin. In cultured macrophages the macromolecular prodrugs exhibited decreased toxicity while maintaining the anti-inflammatory action of ribavirin.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Zinc acrylate, 98%