Merck
  • Macrophage-derived matrix vesicles: an alternative novel mechanism for microcalcification in atherosclerotic plaques.
Macrophage-derived matrix vesicles: an alternative novel mechanism for microcalcification in atherosclerotic plaques.
Circulation research (2013-04-26)
Sophie E P New, Claudia Goettsch, Masanori Aikawa, Julio F Marchini, Manabu Shibasaki, Katsumi Yabusaki, Peter Libby, Catherine M Shanahan, Kevin Croce, Elena Aikawa
ABSTRACT
We previously showed that early calcification of atherosclerotic plaques associates with macrophage accumulation. Chronic renal disease and mineral imbalance accelerate calcification and the subsequent release of matrix vesicles (MVs), precursors of microcalcification. We tested the hypothesis that macrophage-derived MVs contribute directly to microcalcification. Macrophages associated with regions of calcified vesicular structures in human carotid plaques (n=136 patients). In vitro, macrophages released MVs with high calcification and aggregation potential. MVs expressed exosomal markers (CD9 and TSG101) and contained S100A9 and annexin V. Silencing S100A9 in vitro and genetic deficiency in S100A9-/- mice reduced MV calcification, whereas stimulation with S100A9 increased calcification potential. Externalization of phosphatidylserine after Ca/P stimulation and interaction of S100A9 and annexin V indicated that a phosphatidylserine-annexin V-S100A9 membrane complex facilitates hydroxyapatite nucleation within the macrophage-derived MV membrane. Our results support the novel concept that macrophages release calcifying MVs enriched in S100A9 and annexin V, which contribute to accelerated microcalcification in chronic renal disease.
Sigma-Aldrich
Hydroxyapatite, Type I, buffered aqueous suspension
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)