Merck
  • Virulence of the Shiga toxin type 2-expressing Escherichia coli O104:H4 German outbreak isolate in two animal models.
Virulence of the Shiga toxin type 2-expressing Escherichia coli O104:H4 German outbreak isolate in two animal models.
Infection and immunity (2013-02-27)
Tonia Zangari, Angela R Melton-Celsa, Aruna Panda, Nadia Boisen, Mark A Smith, Ivan Tatarov, Louis J De Tolla, James P Nataro, Alison D O'Brien
ABSTRACT
In May 2011, a large food-borne outbreak was traced to an unusual O104:H4 enteroaggregative Escherichia coli (EAEC) strain that produced Shiga toxin (Stx) type 2 (Stx2). We developed a mouse model to study the pathogenesis and treatment for this strain and examined the virulence of the isolate for Dutch belted rabbits. O104:H4 strain C227-11 was gavaged into C57BL/6 mice at 10(9) to 10(11) CFU/animal. The infected animals were then given water with ampicillin (Amp; 5 g/liter) ad libitum. The C227-11-infected, Amp-treated C57BL/6 mice exhibited both morbidity and mortality. Kidneys from mice infected with C227-11 showed acute tubular necrosis, a finding seen in mice infected with typical Stx-producing E. coli. We provided anti-Stx2 antibody after infection and found that all of the antibody-treated mice gained more weight than untreated mice and, in another study, that all of the antibody-treated animals lived, whereas 3/8 phosphate-buffered saline-treated mice died. We further compared the pathogenesis of C227-11 with that of an Stx-negative (Stx(-)) O104:H4 isolate, C734-09, and an Stx2(-) phage-cured derivative of C227-11. Whereas C227-11-infected animals lost weight or gained less weight over the course of infection and died, mice infected with either of the Stx(-) isolates did not lose weight and only one mouse died. When the Stx-positive (Stx(+)) and Stx2(-) O104:H4 strains were compared in rabbits, greater morbidity and mortality were observed in rabbits infected with the Stx2(+) isolates than the Stx2(-) isolates. In conclusion, we describe two animal models for EAEC pathogenesis, and these studies show that Stx2 is responsible for most of the virulence observed in C227-11-infected mice and rabbits.
Product Description
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Ampicillin, analytical standard