Merck
  • Hx-amides: DNA sequence recognition by the fluorescent Hx (p-anisylbenzimidazole)•pyrrole and Hx•imidazole pairings.
Hx-amides: DNA sequence recognition by the fluorescent Hx (p-anisylbenzimidazole)•pyrrole and Hx•imidazole pairings.
Bioorganic & medicinal chemistry letters (2013-02-12)
Vijay Satam, Pravin Patil, Balaji Babu, Matthew Gregory, Michael Bowerman, Mia Savagian, Megan Lee, Samuel Tzou, Kevin Olson, Yang Liu, Joseph Ramos, W David Wilson, John P Bingham, Kostantinos Kiakos, John A Hartley, Moses Lee
ABSTRACT
Hx-amides are fluorescent hybrids of imidazole (I)- and pyrrole (P)-containing polyamides and Hoechst 33258, and they bind in the minor groove of specific DNA sequences. Synthesis and DNA binding studies of HxII (5) complete our studies on the first set of Hx-amides: Hx-I/P-I/P. HxPP (2), HxIP (3) and HxPI (4) were reported earlier. Results from DNase I footprinting, biosensor-SPR, CD and ΔTM studies showed that Hx-amides interacted with DNA via the anti-parallel and stacked, side-by-side motif. Hx was found to mimic the DNA recognition properties of two consecutive pyrrole units (PP) in polyamides. Accordingly, the stacked Hx/PP pairing binds preferentially to two consecutive AT base pairs, A/T-A/T; Hx/IP prefers C-A/T; Hx/PI prefers A/T-C; and Hx/II prefers C-C. The results also showed that Hx-amides bound their cognate sequence at a higher affinity than their formamido-triamide counterparts.
Sigma-Aldrich
Benzimidazole, 98%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Imidazole, European Pharmacopoeia (EP) Reference Standard