Merck
  • Dietary conjugated linoleic acid modify gene expression in liver, muscles, and fat tissues of finishing pigs.
Dietary conjugated linoleic acid modify gene expression in liver, muscles, and fat tissues of finishing pigs.
Journal of animal science (2013-02-13)
N Tous, P K Theil, C Lauridsen, R Lizardo, B Vilà, E Esteve-Garcia
ABSTRACT
The aim of this study was to investigate underlying mechanisms of dietary conjugated linoleic acid (CLA) on lipid metabolism in various tissues of pigs. Sixteen gilts (73 ± 3 kg) were fed a control (containing sunflower oil) or an experimental diet in which 4% of sunflower oil was replaced by CLA, and slaughtered at an average BW of 117 ± 4.9 kg. Transcription of peroxisome proliferator-activated receptor alpha (PPARα), peroxisome proliferator-activated receptor gamma (PPARγ), fatty acid synthase (FAS), sterol regulatory element binding protein (SREBP1), acetyl-CoA carboxylase (ACC), lipoprotein lipase (LPL), delta-6-desaturase (D6D), and stearoyl CoA desaturase (SCD) were determined by real-time PCR in longissimus thoracis (LT) and semimembranosus (SM) muscles, LT subcutaneous and SM intermuscular fat, and in the liver. Fatty acid (FA) composition was analyzed using gas chromatography in these tissues, except for SM intermuscular fat. Dietary CLA increased PPARγ in LT muscle (P < 0.05), whereas CLA reduced PPARα transcription in all tissues studied (P < 0.05) with the exception of intermuscular fat. Transcription of genes related to FA synthesis was reduced by CLA in SM muscle and liver (SREBP1, both P < 0.1; ACC, P < 0.01 in SM; and FAS, P < 0.01 in liver), whereas CLA reduced (P < 0.05) LPL and D6D transcriptions in SM muscle and reduced (P < 0.05) SCD in liver but increased (P < 0.05) SCD in LT muscle and intermuscular fat. Saturated FA were increased in all studied tissues (P < 0.01), while monosaturated and polyunsaturated FA were reduced in a tissue-specific way by CLA. It was concluded that dietary CLA affected transcription of genes and fat metabolism in a tissue-specific manner.
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus
Supelco
Sunflower seed oil, analytical standard