Merck
  • [Iron and zinc in vitro potential availability in an infant diet with fortified bread with different iron sources or with the addition of different iron absorption promoters].
[Iron and zinc in vitro potential availability in an infant diet with fortified bread with different iron sources or with the addition of different iron absorption promoters].
Archivos latinoamericanos de nutricion (2012-06-16)
María J Binaghi, Carolina E Cagnasso, Nestor R Pellegrino, Silvina R Drago, Rolando González, Patricia A Ronayne, Mirta E Valencia
ABSTRACT
Home-made diets are the most frequently used complementary foods. In the present work we evaluated iron and zinc availability in a usually consumed infant diet containing either iron-fortified bread with different iron sources: ferrous sulfate, ferrous bisglycinate, NaFeEDTA. We also used non-fortified bread with absorption promoters: ascorbic acid, sodium citrate, Na2EDTA, combined with different beverages. The diet (potato, pumpkin, grits, bread, and apple) was combined with water, milk, tea, a soft drink and an orange-based artificial drink. Mineral dialyzability (D) as an indicator of potential availability was determined using an in vitro method. Statistical analysis was performed by ANOVA, and a posteriori Tukey test. There were no significant differences in FeD between diets with ferrous sulfate or ferrous bisglycinate fortified bread; in NaFeEDTA fortified bread it increased significantly (p<0.05). Iron D increase was greater in diets with bread containing absorption promoters than in those with fortified bread. The orange-based artificial drink increased FeD, while tea and milk decreased it significantly (p < 0.05). Zinc D increased significantly when the bread was fortified either with ferrous sulfate or NaFeEDTA, but remained unchanged in diets with ferrous bisglycinate fortified bread. The addition of tea or milk decreased ZnD while the orange-based artificial drink increased it significantly (p < 0.05). Regarding absorption promoters, the greater values both in FeD and ZnD were observed in diets with iron nonfortified bread containing Na2EDTA.
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, BioReagent, suitable for plant cell culture, crystalline