Merck
Giant pulmonary and hepatic hydatid cysts in a child.
Tropical doctor (2011-11-08)
Crenguta Livia Calma, Adriana Maria Neghina, Brigitha Vlaicu, Raul Neghina
ABSTRACT
This case report focuses on hydatidosis, one of the most important parasitic zoonoses in Romania. We describe an interesting paediatric case of an eight-year old boy from a rural region presenting with a both giant and life-threatening pulmonary cyst (that was successfully managed) and a hepatic cyst. Noteworthy are the negative consequences of this disorder on children's health and development as a result of prolonged hospitalizations, surgical interventions and chemotherapy.
Product Description
Supelco
Hydrocortisone 21-hemisuccinate, analytical standard
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, powder, BioReagent, suitable for cell culture