Merck
  • An improved palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.
An improved palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.
Organic letters (2011-08-26)
Jun Pan, Xinyan Wang, Yong Zhang, Stephen L Buchwald
ABSTRACT
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with greatly simplified conditions relative to our previous report.
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis