Merck
  • Dechlorination of individual congeners in aroclor 1248 as enhanced by chlorobenzoates, chlorophenols, and chlorobenzenes.
Dechlorination of individual congeners in aroclor 1248 as enhanced by chlorobenzoates, chlorophenols, and chlorobenzenes.
Journal of microbiology and biotechnology (2008-10-29)
Jongseol Kim, Young-Cheol Cho, Robert C Frohnhoefer, G-Yull Rhee
ABSTRACT
Previous investigations showed that three classes of haloaromatic compounds (HACs; chlorobenzoates, chlorophenols, and chlorobenzenes) enhanced the reductive dechlorination of Aroclor 1248, judging from the overall extent of reduction in Cl atoms on the biphenyl. In the present study, we further investigated the kind of polychlorinated biphenyl (PCB) congeners involved in the enhanced dechlorination by four isomers belonging to each class (2,3-, 2,5-, 2,3,5-, and 2,4,6-chlorobenzoates; 2,3-, 3,4-, 2,5-, and 2,3,6-chlorophenols; and 1,2-, 1,2,3-, 1,2,4-, and penta-chlorobenzenes). Although the PCB congeners involved in the enhanced dechlorination varied with the HACs, the enhancement primarily involved paradechlorination of the same congeners (2,3,4'-, 2,3,4,2'- plus 2,3,6,4'-, 2,5,3',4'- plus 2,4,5,2',6'-, and 2,3,6,2',4'- chlorobiphenyls), regardless of the HACs. These congeners are known to have low threshold concentrations for dechlorination. To a lesser extent, the enhancement also involved meta dechlorination of certain congeners with high threshold concentrations. There was no or less accumulation of 2,4,4'- and 2,5,4'-chlorobiphenyls as final products under HAC amendment. Although the dechlorination products varied, the accumulation of orthosubstituted congeners, 2-, 2,2'-, and 2,6-chlorobiphenyls, was significantly higher with the HACs, indicating a more complete dechlorination of the highly chlorinated congeners. Therefore, the present results suggest that the enhanced dechlorination under HAC enrichment is carried out through multiple pathways, some of which may be universal, regardless of the kind of HACs, whereas others may be HAC-specific.
Product Description
Supelco
Aroclor 1248 solution, certified reference material, 1000 μg/mL in isooctane
Pricing and availability is not currently available.