Merck
  • Modified carbon paste sensor for cetyltrimethylammonium ion based on its ion-associate with tetrachloropalladate(II).
Modified carbon paste sensor for cetyltrimethylammonium ion based on its ion-associate with tetrachloropalladate(II).
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2007-05-15)
Hosny Ibrahim, Amal Khorshid
ABSTRACT
A comparative study was made between developed chemically modified carbon paste electrodes and PVC membrane electrodes for the cationic surfactant cetyltrimethylammonium bromide (CTAB). The carbon paste electrode modified with cetyltrimethylammonium-tetrachloropalladate(II) (CTA-TClP) provides a more sensitive and stable device than that shown by electrodes with an inner reference solution. The best performance was obtained by an electrode based on the paste containing 3.6 wt% CTA-TCIP, 1.8 wt% ethylhexadecyldimethylammonium bromide, 37.6 wt% graphite and 57 wt% tricresyl phosphate. The sensor exhibited a Nernstian response for CTAB over a wide concentration range of 3.5 x 10(-7) to 1.0 x 10(-3) M with a detection limit of 2.0 x 10(-7) M between pH 2.7 and 8.2 with a fast response time of <or=15 s. The electrode showed excellent selectivity for CTAB over a large number of ions. Interferences caused by common cationic surfactants have been investigated in simulated mixtures containing high concentration levels of interfering surfactants, and the sensor was found to be tolerant against these compounds.
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%