Merck
  • Biotransformation of D-limonene to (+) trans-carveol by toluene-grown Rhodococcus opacus PWD4 cells.
Biotransformation of D-limonene to (+) trans-carveol by toluene-grown Rhodococcus opacus PWD4 cells.
Applied and environmental microbiology (2001-05-26)
W A Duetz, A H Fjällman, S Ren, C Jourdat, B Witholt
ABSTRACT
The toluene-degrading strain Rhodococcus opacus PWD4 was found to hydroxylate D-limonene exclusively in the 6-position, yielding enantiomerically pure (+) trans-carveol and traces of (+) carvone. This biotransformation was studied using cells cultivated in chemostat culture with toluene as a carbon and energy source. The maximal specific activity of (+) trans-carveol formation was 14.7 U (g of cells [dry weight])(-1), and the final yield was 94 to 97%. Toluene was found to be a strong competitive inhibitor of the D-limonene conversion. Glucose-grown cells did not form any trans-carveol from D-limonene. These results suggest that one of the enzymes involved in toluene degradation is responsible for this allylic monohydroxylation. Another toluene degrader (Rhodococcus globerulus PWD8) had a lower specific activity but was found to oxidize most of the formed trans-carveol to (+) carvone, allowing for the biocatalytic production of this flavor compound.
Sigma-Aldrich
L-Carveol, mixture of cis and trans, ≥95%, FG
Sigma-Aldrich
(−)-Carveol, mixture of isomers, 97%