Merck
  • Mutations in human GLUD2 glutamate dehydrogenase affecting basal activity and regulation.
Mutations in human GLUD2 glutamate dehydrogenase affecting basal activity and regulation.
Journal of neurochemistry (2009-05-07)
Konstantinos Kanavouras, Nikolas Borompokas, Helen Latsoudis, Argiris Stagourakis, Ioannis Zaganas, Andreas Plaitakis
ABSTRACT
Glutamate dehydrogenase (GDH) in human exists in GLUD1 and GLUD2 gene-encoded isoforms (hGDH1 and hGDH2, respectively), differing in their regulation and tissue expression pattern. Whereas hGDH1 is subject to GTP control, hGDH2 uses for its regulation, a novel molecular mechanism not requiring GTP. This is based on the ability of hGDH2 to maintain a baseline activity of <10% of its capacity subject to full activation by rising ADP/L-leucine levels. Here we studied further the molecular mechanisms regulating hGDH2 function by creating and analyzing hGDH2 mutants harboring single amino acid substitutions in the regulatory domain (antenna, pivot helix) of the protein. Five hGDH2 mutants were obtained: two with an amino acid change (Gln441Arg, Ser445Leu) in the antenna, two (Lys450Glu, His454Tyr) in the pivot helix, and one (Ser448Pro) in the junction between the two structures. Functional analyses revealed that, while the antenna mutations increased basal enzyme activity without affecting its allosteric properties, the pivot helix mutations drastically reduced basal activity and impaired enzyme regulation. On the other hand, the Ser448Pro mutation reduced basal activity but did not alter allosteric regulation. Also, compared with wild-type hGDH2, the antenna mutants were relatively thermostable, whereas the pivot helix mutants were extremely heat labile. Hence, the present data further our understanding of the molecular mechanisms involved in the function and stability of hGDH2, an enzyme thought to be of importance for nerve tissue biology.
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type III, lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type II, 50% glycerol solution, ≥35 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type I, ammonium sulfate suspension, ≥40 units/mg protein