Merck
  • An improved Erk biosensor detects oscillatory Erk dynamics driven by mitotic erasure during early development.
An improved Erk biosensor detects oscillatory Erk dynamics driven by mitotic erasure during early development.
Developmental cell (2023-09-16)
Scott G Wilcockson, Luca Guglielmi, Pablo Araguas Rodriguez, Marc Amoyel, Caroline S Hill
ABSTRACT
Extracellular signal-regulated kinase (Erk) signaling dynamics elicit distinct cellular responses in a variety of contexts. The early zebrafish embryo is an ideal model to explore the role of Erk signaling dynamics in vivo, as a gradient of activated diphosphorylated Erk (P-Erk) is induced by fibroblast growth factor (Fgf) signaling at the blastula margin. Here, we describe an improved Erk-specific biosensor, which we term modified Erk kinase translocation reporter (modErk-KTR). We demonstrate the utility of this biosensor in vitro and in developing zebrafish and Drosophila embryos. Moreover, we show that Fgf/Erk signaling is dynamic and coupled to tissue growth during both early zebrafish and Drosophila development. Erk activity is rapidly extinguished just prior to mitosis, which we refer to as mitotic erasure, inducing periods of inactivity, thus providing a source of heterogeneity in an asynchronously dividing tissue. Our modified reporter and transgenic lines represent an important resource for interrogating the role of Erk signaling dynamics in vivo.
Sigma-Aldrich
Cdk1 Inhibitor IV, RO-3306, InSolution, ≥95%
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
MEK1/2 Inhibitor III, PD0325901, InSolution, ≥95%
Sigma-Aldrich
Tyramine hydrochloride, ≥98%