Merck
  • Exercise Training Promotes Sex-Specific Adaptations in Mouse Inguinal White Adipose Tissue.
Exercise Training Promotes Sex-Specific Adaptations in Mouse Inguinal White Adipose Tissue.
Diabetes (2021-02-11)
Pasquale Nigro, Roeland J W Middelbeek, Christiano R R Alves, Susana Rovira-Llopis, Krithika Ramachandran, Leslie A Rowland, Andreas B Møller, Hirokazu Takahashi, Ana B Alves-Wagner, Maria Vamvini, Nathan S Makarewicz, Brent G Albertson, Michael F Hirshman, Laurie J Goodyear
ABSTRACT
Recent studies demonstrate that adaptations to white adipose tissue (WAT) are important components of the beneficial effects of exercise training on metabolic health. Exercise training favorably alters the phenotype of subcutaneous inguinal WAT (iWAT) in male mice, including decreasing fat mass, improving mitochondrial function, inducing beiging, and stimulating the secretion of adipokines. In this study, we find that despite performing more voluntary wheel running compared with males, these adaptations do not occur in the iWAT of female mice. Consistent with sex-specific adaptations, we report that mRNA expression of androgen receptor coactivators is upregulated in iWAT from trained male mice and that testosterone treatment of primary adipocytes derived from the iWAT of male, but not female mice, phenocopies exercise-induced metabolic adaptations. Sex specificity also occurs in the secretome profile, as we identify cysteine-rich secretory protein 1 (Crisp1) as a novel adipokine that is only secreted from male iWAT in response to exercise. Crisp1 expression is upregulated by testosterone and functions to increase glucose and fatty acid uptake. Our finding that adaptations to iWAT with exercise training are dramatically greater in male mice has potential clinical implications for understanding the different metabolic response to exercise training in males and females and demonstrates the importance of investigating both sexes in studies of adipose tissue biology.
Sigma-Aldrich
ChIPAb+ Androgen Receptor- ChIP Validated Antibody and Primer Set, from rabbit, purified by affinity chromatography
Roche
Insulin, human, recombinant (yeast)