Merck
  • Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography-Tandem Mass Spectrometry.
Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography-Tandem Mass Spectrometry.
Journal of agricultural and food chemistry (2018-03-15)
Yongho Shin, Jonghwa Lee, Jiho Lee, Junghak Lee, Eunhye Kim, Kwang-Hyeon Liu, Hye Suk Lee, Jeong-Han Kim
ABSTRACT
A screening method for simultaneous analysis of 379 pesticides in human serum was developed using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Electrospray ionization with positive/negative switching mode of LC-MS/MS was adopted, and scheduled multiple reaction monitoring for each target compound was established. The limit of quantitation was 10 ng/mL for 94.5% of the total pesticides, and the correlation coefficients of calibration were ≥0.990 for 93.9% of the pesticides. For the sample preparation, scaled-down QuEChERS were used. Serum (100 μL) was extracted with acetonitrile (400 μL), partitioned with magnesium sulfate (40 mg) and sodium chloride (10 mg), and the upper layer was used for analysis without further cleanup steps. For the accuracy and precision tests, most of the pesticides showed excellent results in intra- and interday conditions. In the recovery tests at 10, 50, and 250 ng/mL, 85.8-91.8% of all target compounds satisfied the recovery range of 70-120% (relative standard deviation ≤20%).
Product Description
Supelco
Sethoxydim, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benthiavalicarb isopropyl, PESTANAL®, analytical standard
Supelco
Amisulbrom, PESTANAL®, analytical standard
Supelco
Azamethiphos, PESTANAL®, analytical standard
Supelco
Tralomethrin, mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland