Merck
  • Puerarin alleviates coronary heart disease via suppressing inflammation in a rat model.
Puerarin alleviates coronary heart disease via suppressing inflammation in a rat model.
Gene (2020-12-18)
Liangping Zhao, Li Wang, Daimin Zhang, Yuqi Chen, Fulu Jin
ABSTRACT
Puerarin shows inhibitory effects on inflammation in chronic heart failure (CHF), but its efficacy in coronary heart disease (CHD) remained vague. Rat CHD model was constructed, and serum parameters were determined using a blood liquid biochemical analyzer. Also, contents of creatine kinase (CK), creatine kinase MB isoenzyme (CK-MB), lactate dehydrogenase (LDH) and cardiac troponin (cTnT) were measured using colorimetry. Histological examination was conducted with Hematoxylin-Eosin (H&E) staining, and cardiac function was assessed by Echocardiography. Cell apoptosis was detected using Terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling (TUNEL) assay. Relative expressions were measured using quantitative real-time polymerase chain reaction (qRT-PCR) and Western blot as needed. In CHD rats, the levels of TC, LDL and TG and the expressions of matrix metalloproteinase-9 (MMP-9), CD40 ligand (CD40L), tumor necrosis factor-α (TNF-α) and C-reactive protein (CRP) were increased while HDL level was decreased, accompanied with inflammatory cell infiltration and cardiac malfunction. Also, the contents of CK, CK-MB, LDH and cTnT, the percentage of apoptotic cells, the expressions of Bcl-2 associated X protein (Bax), cleaved Caspase-3, TNF-α, Interleukin-β (IL-β), IL-6 and Lipoprotein-associated Phospholipase A2 (Lp-PLA2) expressions and the levels of oxidized-(ox-)LDL and malondialdehyde (MDA) were upregulated, while the level of super oxidase dismutase (SOD) and the expressions of B cell lymphoma-2 (Bcl-2) and vascular endothelial growth factor (VEGF) were downregulated. However, Puerarin ameliorated the effects of CHD model construction, suppressed nuclear factor-(NF-)κB expression, and enhanced the expressions of Farnesoid X Receptor (FXR), phosphorylated-AKT (p-AKT) and phosphorylated-signal transducer and activator of transcription 3 (p-STAT3). Puerarin alleviated CHD in rats via inhibiting inflammation, providing possible method for CHD treatment.
Sigma-Aldrich
Egg yolks from chicken, powder
Sigma-Aldrich
Sodium cholate hydrate, from bovine and/or ovine bile, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Lard oil
Propylthiouracil, European Pharmacopoeia (EP) Reference Standard