Merck
  • An Expanded Surface-Enhanced Raman Scattering Tags Library by Combinatorial Encapsulation of Reporter Molecules in Metal Nanoshells.
An Expanded Surface-Enhanced Raman Scattering Tags Library by Combinatorial Encapsulation of Reporter Molecules in Metal Nanoshells.
ACS nano (2020-09-02)
Sergio Rodal-Cedeira, Alba Vázquez-Arias, Gustavo Bodelón, Alexander Skorikov, Sara Núñez-Sánchez, Andrea Laporta, Lakshminarayana Polavarapu, Sara Bals, Luis M Liz-Marzán, Jorge Pérez-Juste, Isabel Pastoriza-Santos
ABSTRACT
Raman-encoded gold nanoparticles (NPs) have been widely employed as photostable multifunctional probes for sensing, bioimaging, multiplex diagnostics, and surface-enhanced Raman scattering (SERS)-guided tumor therapy. We report a strategy toward obtaining a particularly large library of Au nanocapsules encoded with Raman codes defined by the combination of different thiol-free Raman reporters, encapsulated at defined molar ratios. The fabrication of SERS tags with tailored size and predefined codes is based on the in situ incorporation of Raman reporter molecules inside Au nanocapsules during their formation via galvanic replacement coupled to seeded growth on Ag NPs. The hole-free closed-shell structure of the nanocapsules is confirmed by electron tomography. The unusually wide encoding possibilities of the obtained SERS tags are investigated by means of either wavenumber-based encoding or Raman frequency combined with signal intensity, leading to an outstanding performance as exemplified by 26 and 54 different codes, respectively. We additionally demonstrate that encoded nanocapsules can be readily bioconjugated with antibodies for applications such as SERS-based targeted cell imaging and phenotyping.
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.