Merck
  • Developmental changes in plasticity, synaptic, glia, and connectivity protein levels in rat medial prefrontal cortex.
Developmental changes in plasticity, synaptic, glia, and connectivity protein levels in rat medial prefrontal cortex.
Learning & memory (Cold Spring Harbor, N.Y.) (2018-09-19)
Margaret Jia, Alessio Travaglia, Gabriella Pollonini, Giuseppe Fedele, Cristina M Alberini
ABSTRACT
The medial prefrontal cortex (mPFC) plays a critical role in complex brain functions including decision-making, integration of emotional, and cognitive aspects in memory processing and memory consolidation. Because relatively little is known about the molecular mechanisms underlying its development, we quantified rat mPFC basal expression levels of sets of plasticity, synaptic, glia, and connectivity proteins at different developmental ages. Specifically, we compared the mPFC of rats at postnatal day 17 (PN17), when they are still unable to express long-term contextual and spatial memories, to rat mPFC at PN24, when they have acquired the ability of long-term memory expression and finally to the mPFC of adult rats. We found that, with increased age, there are remarkable and significant decreases in markers of cell activation and significant increases in proteins that mark synaptogenesis and synapse maturation. Furthermore, we found significant changes in structural markers over the ages, suggesting that structural connectivity of the mPFC increases over time. Finally, the substantial biological difference in mPFC at different ages suggest caution in extrapolating conclusions from brain plasticity studies conducted at different developmental stages.
Sigma-Aldrich
Anti-Growth Associated Protein-43 (GAP-43) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-TrkB (Tyr816) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-CaM Kinase II Antibody, α subunit, clone 6G9, clone 6G9, Upstate®, from mouse
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-MBP Antibody, clone SKB3, clone SKB3, Upstate®, from mouse
Sigma-Aldrich
Anti-c-Fos (Ab-2) (4-17) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-GAD65 Antibody, from rabbit, purified by affinity chromatography