Merck
  • Short-chain fatty acids bind to apoptosis-associated speck-like protein to activate inflammasome complex to prevent Salmonella infection.
Short-chain fatty acids bind to apoptosis-associated speck-like protein to activate inflammasome complex to prevent Salmonella infection.
PLoS biology (2020-09-30)
Hitoshi Tsugawa, Yasuaki Kabe, Ayaka Kanai, Yuki Sugiura, Shigeaki Hida, Shun'ichiro Taniguchi, Toshio Takahashi, Hidenori Matsui, Zenta Yasukawa, Hiroyuki Itou, Keiyo Takubo, Hidekazu Suzuki, Kenya Honda, Hiroshi Handa, Makoto Suematsu
ABSTRACT
Short-chain fatty acids (SCFAs) produced by gastrointestinal microbiota regulate immune responses, but host molecular mechanisms remain unknown. Unbiased screening using SCFA-conjugated affinity nanobeads identified apoptosis-associated speck-like protein (ASC), an adaptor protein of inflammasome complex, as a noncanonical SCFA receptor besides GPRs. SCFAs promoted inflammasome activation in macrophages by binding to its ASC PYRIN domain. Activated inflammasome suppressed survival of Salmonella enterica serovar Typhimurium (S. Typhimurium) in macrophages by pyroptosis and facilitated neutrophil recruitment to promote bacterial elimination and thus inhibit systemic dissemination in the host. Administration of SCFAs or dietary fibers, which are fermented to SCFAs by gut bacteria, significantly prolonged the survival of S. Typhimurium-infected mice through ASC-mediated inflammasome activation. SCFAs penetrated into the inflammatory region of the infected gut mucosa to protect against infection. This study provided evidence that SCFAs suppress Salmonella infection via inflammasome activation, shedding new light on the therapeutic activity of dietary fiber.
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution