Merck
  • A Clickable APEX Probe for Proximity-Dependent Proteomic Profiling in Yeast.
A Clickable APEX Probe for Proximity-Dependent Proteomic Profiling in Yeast.
Cell chemical biology (2020-05-30)
Yi Li, Caiping Tian, Keke Liu, Ying Zhou, Jing Yang, Peng Zou
ABSTRACT
The engineered ascorbate peroxidase (APEX) is a powerful tool for the proximity-dependent labeling of proteins and RNAs in live cells. Although widely use in mammalian cells, APEX applications in microorganisms have been hampered by the poor labeling efficiency of its biotin-phenol (BP) substrate. In this study, we sought to address this challenge by designing and screening a panel of alkyne-functionalized substrates. Our best probe, Alk-Ph, substantially improves APEX-labeling efficiency in intact yeast cells, as it is more cell wall-permeant than BP. Through a combination of protein-centric and peptide-centric chemoproteomic experiments, we have identified 165 proteins with a specificity of 94% in the yeast mitochondrial matrix. In addition, we have demonstrated that Alk-Ph is useful for proximity-dependent RNA labeling in yeast, thus expanding the scope of APEX-seq. We envision that this improved APEX-labeling strategy would set the stage for the large-scale mapping of spatial proteome and transcriptome in yeast.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
ORBO 47 Amberlite XAD®-7 specially treated, 100/50 mg, W,W,W separators, O.D. × L 6 mm × 90 mm, pkg of 50 ea
Sigma-Aldrich
Cy5-azide, 95% (HPLC)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
4-(2-Aminoethyl)aniline, 97%
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid