Merck
Structure of mixed micelles formed in PEG-lipid/lipid dispersions.
Langmuir : the ACS journal of surfaces and colloids (2007-03-09)
Maria C Sandström, Emma Johansson, Katarina Edwards
ABSTRACT
Polyethylene glycol (PEG)-conjugated lipids are commonly employed for steric stabilization of liposomes. When added in high concentrations PEG-lipids induce formation of mixed micelles, and depending on the lipid composition of the sample, these may adapt either a discoidal or a long threadlike shape. The factors governing the type of micellar aggregate formed have so far not been investigated in detail. In this study we have systematically varied the lipid composition in lipid/PEG-lipid mixtures and characterized the aggregate structure by means of cryo-transmission electron microscopy (cryo-TEM). The effects caused by adding sterols, phosphatidylethanolamines, and phospholipids with saturated acyl chains to egg phosphatidylcholine/1,2-distearoyl-sn-glycero-3-phosphatidylethanolamine-N-[methoxy(polyethylene glycol)-2000 (EPC/DSPE-PEG2000) mixtures with a fixed amount (25 mol %) of DSPE-PEG2000 was studied. Further, the aggregate structure in 1,2-dimyristoyl-sn-glycero-3-phosphatidylcholine/1,2-dimyristoyl-sn-glycero-3-phosphatidylethanolamine-N-[methoxy(polyethylene glycol)-2000] (DMPC/DMPE-PEG2000) samples above and below the gel to liquid crystalline phase transition temperature (TC) was investigated. Our results revealed that lipid components, as well as environmental conditions, that reduce the lipid spontaneous curvature and increase the monolayer bending modulus tend to promote formation of discoidal micelles. At temperatures below the gel-to-liquid crystalline phase transition temperature reduced lipid/PEG-lipid miscibility, furthermore, likely contribute to the observed formation of discoidal rather than threadlike micelles.
Product Description
Avanti
18:1 PEG350 PE, Avanti Research - A Croda Brand 880430O
Avanti
18:1 PEG1000 PE, Avanti Research - A Croda Brand 880730C
Avanti
18:1 PEG750 PE, Avanti Research - A Croda Brand 880630O
Avanti
18:1 PEG1000 PE, Avanti Research - A Croda Brand 880730P, powder
Avanti
16:0 PEG550 PE, Avanti Research - A Croda Brand 880500P, powder
Avanti
16:0 PEG350 PE, Avanti Research - A Croda Brand 880400P, powder
Avanti
18:1 PEG550 PE, Avanti Research - A Croda Brand 880530C
Avanti
18:1 PEG550 PE, Avanti Research - A Croda Brand 880530O
Avanti
16:0 PEG750 PE, Avanti Research - A Croda Brand 880600P, powder