Merck
  • Evidence for antibody-mediated pathogenesis in anti-NMDAR encephalitis associated with ovarian teratoma.
Evidence for antibody-mediated pathogenesis in anti-NMDAR encephalitis associated with ovarian teratoma.
Acta neuropathologica (2009-08-15)
Erdem Tüzün, Lei Zhou, Joachim M Baehring, Serguei Bannykh, Myrna R Rosenfeld, Josep Dalmau
ABSTRACT
We report the immunopathological analysis of the brain and tumor of two patients who died of anti-NMDAR-associated encephalitis, and of the tumor of nine patients who recovered. Findings included prominent microgliosis and deposits of IgG with rare inflammatory infiltrates in the hippocampus, forebrain, basal ganglia, and spinal cord. Detection of cells expressing markers of cytotoxicity (TIA, granzyme B, perforin and Fas/Fas ligand) was extremely uncommon. All tumors showed NMDAR-expressing neurons and inflammatory infiltrates. All patients’ NMDAR antibodies were IgG1, IgG2, or IgG3. No complement deposits were observed in any of the central nervous system regions examined. Overall, these findings coupled with recently reported in vitro data showing that antibodies downregulate the levels of NMDA receptors suggest that the antibody immune-response is more relevant than cytotoxic T-cell mechanisms in the pathogenesis of anti-NMDAR-associated encephalitis.
Sigma-Aldrich
Anti-Human IgG4−FITC antibody, Mouse monoclonal, clone HP-6025, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG3−FITC antibody, Mouse monoclonal, clone HP-6050, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG2−FITC antibody, Mouse monoclonal, clone HP-6014, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG1−FITC antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture