Merck
mTOR pathway activation drives lung cell senescence and emphysema.
JCI insight (2018-02-09)
Amal Houssaini, Marielle Breau, Kanny Kebe, Shariq Abid, Elisabeth Marcos, Larissa Lipskaia, Dominique Rideau, Aurelien Parpaleix, Jin Huang, Valerie Amsellem, Nora Vienney, Pierre Validire, Bernard Maitre, Aya Attwe, Christina Lukas, David Vindrieux, Jorge Boczkowski, Genevieve Derumeaux, Mario Pende, David Bernard, Silke Meiners, Serge Adnot
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is a highly prevalent and devastating condition for which no curative treatment is available. Exaggerated lung cell senescence may be a major pathogenic factor. Here, we investigated the potential role for mTOR signaling in lung cell senescence and alterations in COPD using lung tissue and derived cultured cells from patients with COPD and from age- and sex-matched control smokers. Cell senescence in COPD was linked to mTOR activation, and mTOR inhibition by low-dose rapamycin prevented cell senescence and inhibited the proinflammatory senescence-associated secretory phenotype. To explore whether mTOR activation was a causal pathogenic factor, we developed transgenic mice exhibiting mTOR overactivity in lung vascular cells or alveolar epithelial cells. In this model, mTOR activation was sufficient to induce lung cell senescence and to mimic COPD lung alterations, with the rapid development of lung emphysema, pulmonary hypertension, and inflammation. These findings support a causal relationship between mTOR activation, lung cell senescence, and lung alterations in COPD, thereby identifying the mTOR pathway as a potentially new therapeutic target in COPD.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests